Learn More
Waters Corp ACQUITY UPLC VanGuard FIT Cartridge, 1.7 μm, 2.1 mm ID, 5 mm
Catalog No. 502039065 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY Premier cartridges are designed to work with ACQUITY Premier VanGuard FIT columns. These cartridges provide fully integrated guard protection for the analytical column, shielding it from matrix and sample contaminants, which helps to extend the column's lifetime.
  • Works with VanGuard FIT columns
  • Fully integrated guard cartridges
  • Protects the analytical column from matrix and sample contaminants
  • Extends column lifetime
Catalog No. 50-203-9065 Supplier Waters Corp Supplier No. 186009459
ACQUITY Premier cartridges are designed to work with ACQUITY Premier VanGuard FIT columns. These cartridges provide fully integrated guard protection for the analytical column, shielding it from matrix and sample contaminants, which helps to extend the column's lifetime.
  • Works with VanGuard FIT columns
  • Fully integrated guard cartridges
  • Protects the analytical column from matrix and sample contaminants
  • Extends column lifetime