Learn More
Waters Corp Waters Column, C8, 1.7 μm, 2.1 mm x 100 mm
Catalog No. 502269365 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY Premier BEH C8 Columns offer less retention than C18 Columns, ideal for fast method development. They feature trifunctionally bonded BEH particles, providing a wide pH range (1 to 12), superior low pH stability, and ultra-low column bleed. MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • Less retentive compared to BEH C18 columns, which is desirable for fast methods development.
  • Trifunctionally bonded BEH particles offer the widest usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed.
  • MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions that can lead to poor peak shape and losses in signal intensity.
This item is not returnable. View return policy
Catalog No. 50-226-9365 Supplier Waters Corp Supplier No. 186010357
ACQUITY Premier BEH C8 Columns offer less retention than C18 Columns, ideal for fast method development. They feature trifunctionally bonded BEH particles, providing a wide pH range (1 to 12), superior low pH stability, and ultra-low column bleed. MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • Less retentive compared to BEH C18 columns, which is desirable for fast methods development.
  • Trifunctionally bonded BEH particles offer the widest usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed.
  • MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions that can lead to poor peak shape and losses in signal intensity.