Learn More
Waters Corp Waters Column, BEH C8, 1.7 μm, 2.1 x 100 mm, VanGuard FIT
Catalog No. 502269368 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
BEH Technology columns are a good universal choice for a wide variety of compounds, offering exceptional peak shape for basic analytes at elevated pH, stability across a wide pH range, and high temperatures. These C8 columns are designed for fast methods development and offer less retention compared to C18 columns.
  • Trifunctionally bonded BEH particles provide a wide usable pH range (1 to 12) and superior low pH stability.
  • Ultra-low column bleed.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions.
  • Minimizes loss of metal-sensitive analytes, including lipids, organic acids, acidic peptides, and oligonucleotides.
  • Integrated VanGuard Fit column design protects the column from contaminants and extends its lifetime.
  • Improved sensitivity and peak shapes.
  • Reduced column conditioning and passivation times.
This item is not returnable. View return policy
Catalog No. 50-226-9368 Supplier Waters Corp Supplier No. 186010360
BEH Technology columns are a good universal choice for a wide variety of compounds, offering exceptional peak shape for basic analytes at elevated pH, stability across a wide pH range, and high temperatures. These C8 columns are designed for fast methods development and offer less retention compared to C18 columns.
  • Trifunctionally bonded BEH particles provide a wide usable pH range (1 to 12) and superior low pH stability.
  • Ultra-low column bleed.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions.
  • Minimizes loss of metal-sensitive analytes, including lipids, organic acids, acidic peptides, and oligonucleotides.
  • Integrated VanGuard Fit column design protects the column from contaminants and extends its lifetime.
  • Improved sensitivity and peak shapes.
  • Reduced column conditioning and passivation times.