Learn More
Waters Corp Waters ACQUITY Premier BEH HILIC Column, 1.7 μm, 2.1 mm × 100 mm
Catalog No. 502269386 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY Premier BEH HILIC Columns use unbonded BEH (Bridged Ethyl Hybrid) particles, designed to retain and separate very polar compounds. They are optimized for efficient, reproducible separations across a broad pH range and offer long column lifetimes due to the BEH particle's chemical stability. MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • Designed to retain and separate very polar compounds using unbonded hybrid particles.
  • Optimized for efficient, reproducible separations across a broad pH range.
  • Extended column lifetimes due to chemically stable particles.
  • More rugged than silica-based HILIC phases.
  • Features high performance surface hardware technology.
  • Reduces analyte/surface interactions, improving peak shape and signal intensity.
This item is not returnable. View return policy
Catalog No. 50-226-9386 Supplier Waters Corp Supplier No. 186010378
The ACQUITY Premier BEH HILIC Columns use unbonded BEH (Bridged Ethyl Hybrid) particles, designed to retain and separate very polar compounds. They are optimized for efficient, reproducible separations across a broad pH range and offer long column lifetimes due to the BEH particle's chemical stability. MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • Designed to retain and separate very polar compounds using unbonded hybrid particles.
  • Optimized for efficient, reproducible separations across a broad pH range.
  • Extended column lifetimes due to chemically stable particles.
  • More rugged than silica-based HILIC phases.
  • Features high performance surface hardware technology.
  • Reduces analyte/surface interactions, improving peak shape and signal intensity.