Learn More
Waters Corp Waters ACQUITY Premier BEH HILIC Column, 1.7 μm, 2.1 mm x 50 mm
Catalog No. 502269385 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY Premier BEH HILIC Columns are designed with unbonded BEH (Bridged Ethyl Hybrid) particles to effectively retain and separate highly polar compounds. These columns ensure efficient and reproducible separations across a broad pH range, offering enhanced ruggedness and extended lifetimes compared to silica-based HILIC phases.
  • Optimized for efficient and reproducible separations across a wide pH range.
  • Features BEH particle chemical stability for long column lifetimes.
  • Incorporates MaxPeak High Performance Surface hardware technology.
  • Reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
This item is not returnable. View return policy
Catalog No. 50-226-9385 Supplier Waters Corp Supplier No. 186010377
ACQUITY Premier BEH HILIC Columns are designed with unbonded BEH (Bridged Ethyl Hybrid) particles to effectively retain and separate highly polar compounds. These columns ensure efficient and reproducible separations across a broad pH range, offering enhanced ruggedness and extended lifetimes compared to silica-based HILIC phases.
  • Optimized for efficient and reproducible separations across a wide pH range.
  • Features BEH particle chemical stability for long column lifetimes.
  • Incorporates MaxPeak High Performance Surface hardware technology.
  • Reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.