Learn More
Waters Corp Waters ACQUITY Premier BEH HILIC Column, 1.7 μm, 2.1 mm x 150 mm, 130 Å
Catalog No. 502269387 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters ACQUITY Premier BEH HILIC Columns are engineered with unbonded BEH (Bridged Ethyl Hybrid) particles, ideal for separating highly polar compounds. They offer efficient, reproducible separations across diverse pH ranges, boasting enhanced ruggedness and chemical stability for extended column lifetimes. Integrated MaxPeak High Performance Surface technology minimizes analyte-surface interactions, improving peak shape and signal intensity.
  • Separates highly polar compounds.
  • Efficient, reproducible separations across wide pH ranges.
  • Enhanced ruggedness and chemical stability for long column lifetimes.
  • MaxPeak High Performance Surface technology prevents poor peak shape and signal loss.
This item is not returnable. View return policy
Catalog No. 50-226-9387 Supplier Waters Corp Supplier No. 186010379
Waters ACQUITY Premier BEH HILIC Columns are engineered with unbonded BEH (Bridged Ethyl Hybrid) particles, ideal for separating highly polar compounds. They offer efficient, reproducible separations across diverse pH ranges, boasting enhanced ruggedness and chemical stability for extended column lifetimes. Integrated MaxPeak High Performance Surface technology minimizes analyte-surface interactions, improving peak shape and signal intensity.
  • Separates highly polar compounds.
  • Efficient, reproducible separations across wide pH ranges.
  • Enhanced ruggedness and chemical stability for long column lifetimes.
  • MaxPeak High Performance Surface technology prevents poor peak shape and signal loss.