Learn More
Waters Corp MassTrak Immunosuppressant Internal Standard Set
Catalog No. 502269342 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The MassTrak Immunosuppressant Internal Standard is intended for in vitro diagnostic use to account for sample matrix effects in quantitative LC-MS/MS assays for the measurement of immunosuppressants in human specimens.The Waters MassTrak Immunosuppressant Internal Standard Set consists of an internal standard and labeling.Contents include:■ Internal Standard (1 vial)■ Insert Card referencing the electronic IFU (Instructions for Use), SDS (Safety Data Sheets), and COA (Certificate of Analysis)The Internal Standard contains the following analytes:■ Ascomycin■ [13C2 2H4]-Everolimus■ [2H12]-Cyclosporin A
This item is not returnable. View return policy
Catalog No. 50-226-9342 Supplier Waters Corp Supplier No. 186010253IVD
Add to Cart
Add to Cart
The MassTrak Immunosuppressant Internal Standard is intended for in vitro diagnostic use to account for sample matrix effects in quantitative LC-MS/MS assays for the measurement of immunosuppressants in human specimens.The Waters MassTrak Immunosuppressant Internal Standard Set consists of an internal standard and labeling.Contents include:■ Internal Standard (1 vial)■ Insert Card referencing the electronic IFU (Instructions for Use), SDS (Safety Data Sheets), and COA (Certificate of Analysis)The Internal Standard contains the following analytes:■ Ascomycin■ [13C2 2H4]-Everolimus■ [2H12]-Cyclosporin A