Learn More
Waters Corp Waters ACQUITY Premier Column, BEH Phenyl, 1.7 um, 2.1 x 100 mm
Catalog No. 502269347 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY Premier BEH Phenyl Columns offer complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds, ensuring excellent peak shape and long column lifetimes.
  • Complementary selectivity to C18 phases, ideal for polyaromatic compounds
  • Chemically stable phenyl sorbent; ultra-low bleed, long lifetimes, excellent peak shape
  • MaxPeak High Performance Surface hardware reduces unwanted analyte/surface interactions
  • VanGuard Fit design protects against contaminants and extends column life
  • Universal column choice for various compounds
  • Exceptional peak shape for basic analytes at elevated pH
  • Stable across wide pH range and high temperatures
This item is not returnable. View return policy
Catalog No. 50-226-9347 Supplier Waters Corp Supplier No. 186010339
ACQUITY Premier BEH Phenyl Columns offer complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds, ensuring excellent peak shape and long column lifetimes.
  • Complementary selectivity to C18 phases, ideal for polyaromatic compounds
  • Chemically stable phenyl sorbent; ultra-low bleed, long lifetimes, excellent peak shape
  • MaxPeak High Performance Surface hardware reduces unwanted analyte/surface interactions
  • VanGuard Fit design protects against contaminants and extends column life
  • Universal column choice for various compounds
  • Exceptional peak shape for basic analytes at elevated pH
  • Stable across wide pH range and high temperatures