Learn More
Waters Corp Waters ACQUITY Premier BEH Shield RP18 Column, 1.7 μm, 2.1 mm ID, 100 mm Length
Supplier:  Waters Corp 186009498
Encompass_Preferred
ACQUITY Premier BEH Shield RP18 Columns complement ACQUITY Premier phases, offering a robust tool for method development. An embedded carbamate group provides alternative selectivity, particularly for phenolic compounds, compared to traditional straight-chain alkyl columns. Hardware technology significantly reduces unwanted analyte/surface interactions, improving peak shape and signal intensity. This chemistry provides complementary selectivity to C18 columns while enhancing peak shape for basic chemicals and maintaining 100% aqueous mobile phase compatibility.
  • C18 chemistry
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range 2 to 11
  • Maximum pressure 18000 psi (1240 Bar)
  • 1.7 μm particle size
  • 2.1 mm inner diameter
  • 100 mm length
  • 130 Å pore size
  • UPLC system compatible
  • LC technique
Catalog No. 50-203-9098