Learn More
Waters Corp Waters Column, BEH Shield RP18, 1.7 μm, 2.1 x 100 mm
Catalog No. 502039101 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY Premier BEH Shield RP18 Columns are designed to complement ACQUITY Premier phases and serve as a powerful tool for method development. The embedded carbamate group in the bonded phase ligand offers alternate selectivity, particularly for phenolic compounds, compared to straight chain alkyl columns. The MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions, which can cause poor peak shape and signal intensity losses. The VanGuard FIT Column design protects the column from contaminants, thereby increasing its lifetime.
  • Embedded carbamate group in bonded phase ligand for alternate selectivity, especially for phenolic compounds.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions.
  • VanGuard FIT Column design protects the column from contaminants and extends column lifetime.
Catalog No. 50-203-9101 Supplier Waters Corp Supplier No. 186009501
ACQUITY Premier BEH Shield RP18 Columns are designed to complement ACQUITY Premier phases and serve as a powerful tool for method development. The embedded carbamate group in the bonded phase ligand offers alternate selectivity, particularly for phenolic compounds, compared to straight chain alkyl columns. The MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions, which can cause poor peak shape and signal intensity losses. The VanGuard FIT Column design protects the column from contaminants, thereby increasing its lifetime.
  • Embedded carbamate group in bonded phase ligand for alternate selectivity, especially for phenolic compounds.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions.
  • VanGuard FIT Column design protects the column from contaminants and extends column lifetime.