Learn More
Waters Corp Waters Column, BEH Shield RP18, 1.7 μm, 2.1 mm ID, 50 mm Length
Supplier:  Waters Corp 186009497
Encompass_Preferred
ACQUITY Premier BEH Shield RP18 Columns offer an alternate selectivity for method development, particularly for phenolic compounds, due to an embedded carbamate group in the bonded phase ligand. The MaxPeak High Performance Surface hardware technology aims to reduce unwanted analyte/surface interactions, which can cause poor peak shape and signal intensity loss.
  • Alternate selectivity for phenolic compounds compared to straight chain alkyl columns.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions.
  • Embedded carbamate group in the bonded phase ligand.
Catalog No. 50-203-9097