Learn More
Waters Corp Waters Column, C18, 300 Å, 1.7 μm, 2.1 x 50 mm
Catalog No. NC2303894 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is designed for the separation of oligonucleotides using ion-pairing reversed-phase (IP-RP) chromatography on UPLC systems. It features BEH hybrid particle technology for high pH and temperature stability, and MaxPeak Premier high-performance surface technology to minimize non-specific adsorption.
  • Designed for the separation of oligonucleotides using ion-pairing reversed-phase (IP-RP) chromatography on UPLC systems.
  • Features BEH hybrid particle technology for high pH and temperature stability.
  • MaxPeak Premier high-performance surface technology minimizes non-specific adsorption.
  • Each batch of stationary phase is QC tested to ensure consistent performance.
  • Offers high sensitivity, reproducibility, and out-of-the-box performance.
  • 300Å pore size columns provide improved resolution for longer oligonucleotides.
Catalog No. NC2303894 Supplier Waters Corp Supplier No. 186010539
This column is designed for the separation of oligonucleotides using ion-pairing reversed-phase (IP-RP) chromatography on UPLC systems. It features BEH hybrid particle technology for high pH and temperature stability, and MaxPeak Premier high-performance surface technology to minimize non-specific adsorption.
  • Designed for the separation of oligonucleotides using ion-pairing reversed-phase (IP-RP) chromatography on UPLC systems.
  • Features BEH hybrid particle technology for high pH and temperature stability.
  • MaxPeak Premier high-performance surface technology minimizes non-specific adsorption.
  • Each batch of stationary phase is QC tested to ensure consistent performance.
  • Offers high sensitivity, reproducibility, and out-of-the-box performance.
  • 300Å pore size columns provide improved resolution for longer oligonucleotides.