Learn More
Waters Corp Column, CSH C18, 130Å, 1.7 μm, VanGuard FIT, 2.1 x 100 mm, MVK
Catalog No. 502391327 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit contains three columns, each with a different lot. ACQUITY Premier CSH C18 columns address peak shape asymmetry and poor loading capacity for basic compounds in low ionic strength mobile phases. Surface charge enables fast column equilibration between low and high pH mobile phases. MaxPeak High Performance Surface hardware minimizes unwanted analyte/surface interactions, preventing poor peak shape and signal loss. VanGuard Fit design protects against contaminants, extending column lifetime.
  • Addresses peak shape asymmetry & poor loading capacity for basic compounds
  • Enables fast column equilibration for varying pH mobile phases
  • Reduces analyte/surface interactions via MaxPeak High Performance Surface
  • Prevents poor peak shape & signal loss
  • VanGuard Fit protects against contaminants & extends column life
Catalog No. 50-239-1327 Supplier Waters Corp Supplier No. 186009630
This method validation kit contains three columns, each with a different lot. ACQUITY Premier CSH C18 columns address peak shape asymmetry and poor loading capacity for basic compounds in low ionic strength mobile phases. Surface charge enables fast column equilibration between low and high pH mobile phases. MaxPeak High Performance Surface hardware minimizes unwanted analyte/surface interactions, preventing poor peak shape and signal loss. VanGuard Fit design protects against contaminants, extending column lifetime.
  • Addresses peak shape asymmetry & poor loading capacity for basic compounds
  • Enables fast column equilibration for varying pH mobile phases
  • Reduces analyte/surface interactions via MaxPeak High Performance Surface
  • Prevents poor peak shape & signal loss
  • VanGuard Fit protects against contaminants & extends column life