Learn More
Waters Corp ACQUITY Premier CSH C18 Column, 1.7 μm, 130Å, 2.1 x 150 mm, Method Validation Kit
Catalog No. 502392808 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit includes three columns, each packed with a different lot of material. These columns are designed to overcome peak shape asymmetry and poor loading capacity for basic compounds. They feature surface charge for rapid equilibration between low and high pH mobile phases, and MaxPeak High Performance Surface hardware to reduce analyte/surface interactions. The VanGuard Fit design protects the column and extends its lifetime.
  • Method validation kit contains three columns, each with a different lot of material.
  • Overcomes peak shape asymmetry and poor loading capacity for basic compounds.
  • Enables fast column equilibration between low and high pH mobile phases.
  • MaxPeak high performance surface hardware reduces unwanted analyte/surface interactions.
  • VanGuard Fit column design protects the column from contaminants and increases column lifetime.
Catalog No. 50-239-2808 Supplier Waters Corp Supplier No. 186009631
This method validation kit includes three columns, each packed with a different lot of material. These columns are designed to overcome peak shape asymmetry and poor loading capacity for basic compounds. They feature surface charge for rapid equilibration between low and high pH mobile phases, and MaxPeak High Performance Surface hardware to reduce analyte/surface interactions. The VanGuard Fit design protects the column and extends its lifetime.
  • Method validation kit contains three columns, each with a different lot of material.
  • Overcomes peak shape asymmetry and poor loading capacity for basic compounds.
  • Enables fast column equilibration between low and high pH mobile phases.
  • MaxPeak high performance surface hardware reduces unwanted analyte/surface interactions.
  • VanGuard Fit column design protects the column from contaminants and increases column lifetime.