Learn More
Waters Corp Waters Method Validation Kit, CSH C18, 1.7 μm, 2.1 x 100 mm, 130 Å
Catalog No. 502039112 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit contains three columns, designed to improve peak shape and loading capacity for basic compounds, and enable fast column equilibration. The hardware technology reduces unwanted analyte/surface interactions, preventing poor peak shape and signal intensity losses.
  • Overcome peak shape asymmetry and poor loading capacity for basic compounds when using low ionic strength mobile phase.
  • Enable fast column equilibration when switching from low to high pH mobile phases.
  • Hardware technology significantly reduces unwanted analyte/surface interactions.
  • Reduces unwanted analyte/surface interactions that can lead to poor peak shape.
  • Minimizes losses in signal intensity due to surface interactions.
Catalog No. 50-203-9112 Supplier Waters Corp Supplier No. 186009627
This method validation kit contains three columns, designed to improve peak shape and loading capacity for basic compounds, and enable fast column equilibration. The hardware technology reduces unwanted analyte/surface interactions, preventing poor peak shape and signal intensity losses.
  • Overcome peak shape asymmetry and poor loading capacity for basic compounds when using low ionic strength mobile phase.
  • Enable fast column equilibration when switching from low to high pH mobile phases.
  • Hardware technology significantly reduces unwanted analyte/surface interactions.
  • Reduces unwanted analyte/surface interactions that can lead to poor peak shape.
  • Minimizes losses in signal intensity due to surface interactions.