Learn More
Waters Corp ACQUITY Premier CSH C18 Column, 1.7 μm, 2.1 x 50 mm, VanGuard FIT
Catalog No. 502391374 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY Premier CSH C18 Columns address peak shape asymmetry and poor loading capacity for basic compounds in low ionic strength mobile phases. The surface charge aids fast column equilibration between low and high pH mobile phases. MaxPeak High Performance Surface technology minimizes unwanted analyte/surface interactions, improving peak shape and signal intensity. The VanGuard FIT Column design protects the column from contaminants, extending its lifetime.
  • Addresses peak shape asymmetry and poor loading capacity for basic compounds.
  • Aids fast column equilibration (low to high pH).
  • MaxPeak HPS minimizes unwanted analyte/surface interactions.
  • Improves peak shape and signal intensity.
  • Reduces sample losses, mitigates metal-sensitive analyte loss.
  • Enhances sensitivity and recovery.
  • VanGuard FIT design protects column from contaminants.
  • Extends column lifetime.
Catalog No. 50-239-1374 Supplier Waters Corp Supplier No. 186009463
ACQUITY Premier CSH C18 Columns address peak shape asymmetry and poor loading capacity for basic compounds in low ionic strength mobile phases. The surface charge aids fast column equilibration between low and high pH mobile phases. MaxPeak High Performance Surface technology minimizes unwanted analyte/surface interactions, improving peak shape and signal intensity. The VanGuard FIT Column design protects the column from contaminants, extending its lifetime.
  • Addresses peak shape asymmetry and poor loading capacity for basic compounds.
  • Aids fast column equilibration (low to high pH).
  • MaxPeak HPS minimizes unwanted analyte/surface interactions.
  • Improves peak shape and signal intensity.
  • Reduces sample losses, mitigates metal-sensitive analyte loss.
  • Enhances sensitivity and recovery.
  • VanGuard FIT design protects column from contaminants.
  • Extends column lifetime.