Learn More
Waters Corp ACQUITY Column, C18, 1.7 μm, 2.1 mm, 150 mm
Supplier:  Waters Corp 186009462
Encompass_Preferred
ACQUITY Premier CSH C18 Columns address peak shape asymmetry and poor loading capacity for basic compounds in low ionic strength mobile phase. MaxPeak High Performance Surface technology reduces unwanted analyte/surface interactions, improving peak shape and signal. This provides TFA-like peak performance without using TFA, enabling MS-friendly mobile phases and simpler method development.
  • Overcomes peak shape asymmetry and poor loading capacity for basic compounds.
  • MaxPeak High Performance Surface reduces unwanted analyte/surface interactions.
  • Ensures high performance with improved peak shape and less column bleed.
  • Designed for global markets, enabling replicable procedures.
  • Offers maximum loading of basic chemicals under volatile, acidic, low pH conditions.
  • Improved loadability decreases solvent usage and fraction quantities.
Catalog No. 50-203-9068