Learn More
Waters Corp ACQUITY Premier CSH C18 Column, 1.7 μm, 2.1 x 50 mm
Catalog No. 502039066 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY Premier CSH C18 Columns address peak shape asymmetry and limited loading capacity for basic compounds when utilizing low ionic strength mobile phase. The column's surface charge facilitates rapid equilibration when transitioning between low and high pH mobile phases. MaxPeak High Performance Surface hardware technology significantly minimizes undesirable analyte/surface interactions, which can lead to poor peak shape and reduced signal intensity.
  • C18 chemistry, Reversed Phase, Hybrid Spherical particles (1.7 μm, 130 Å).
  • pH range 1 to 11, Max 18000 psi (1240 Bar).
  • Dimensions: 2.1 mm ID x 50 mm L, 15 % Carbon Load.
  • Endcapped, Low Silanol, Parker-style, UPLC, LC, USP L1.
  • eCord included, 1 unit/pkg.
Catalog No. 50-203-9066 Supplier Waters Corp Supplier No. 186009460
ACQUITY Premier CSH C18 Columns address peak shape asymmetry and limited loading capacity for basic compounds when utilizing low ionic strength mobile phase. The column's surface charge facilitates rapid equilibration when transitioning between low and high pH mobile phases. MaxPeak High Performance Surface hardware technology significantly minimizes undesirable analyte/surface interactions, which can lead to poor peak shape and reduced signal intensity.
  • C18 chemistry, Reversed Phase, Hybrid Spherical particles (1.7 μm, 130 Å).
  • pH range 1 to 11, Max 18000 psi (1240 Bar).
  • Dimensions: 2.1 mm ID x 50 mm L, 15 % Carbon Load.
  • Endcapped, Low Silanol, Parker-style, UPLC, LC, USP L1.
  • eCord included, 1 unit/pkg.