Learn More
Waters Corp Waters ACQUITY Premier CSH C18 Column, 1.7 μm, 2.1 mm, 150 mm
Catalog No. NC2245906 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY Premier CSH C18 Columns address peak shape asymmetry and poor loading capacity for basic compounds in low ionic strength mobile phases. Surface charge enables fast column equilibration when switching pH mobile phases. MaxPeak High Performance Surface hardware minimizes unwanted analyte/surface interactions, preventing poor peak shape and reduced signal intensity. VanGuard FIT design protects from contaminants and extends column lifetime.
  • Addresses peak shape asymmetry & poor loading capacity.
  • Enables fast column equilibration.
  • Minimizes unwanted analyte/surface interactions.
  • Extends column lifetime.
  • Improves resolution, sensitivity, and analysis speed.
This item is not returnable. View return policy
Catalog No. NC2245906 Supplier Waters Corp Supplier No. 186009465
ACQUITY Premier CSH C18 Columns address peak shape asymmetry and poor loading capacity for basic compounds in low ionic strength mobile phases. Surface charge enables fast column equilibration when switching pH mobile phases. MaxPeak High Performance Surface hardware minimizes unwanted analyte/surface interactions, preventing poor peak shape and reduced signal intensity. VanGuard FIT design protects from contaminants and extends column lifetime.
  • Addresses peak shape asymmetry & poor loading capacity.
  • Enables fast column equilibration.
  • Minimizes unwanted analyte/surface interactions.
  • Extends column lifetime.
  • Improves resolution, sensitivity, and analysis speed.