Learn More
Waters Corp Waters Column, C18, 1.7 μm, 2.1 mm ID, 100 mm Length
Catalog No. 502039070 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY Premier CSH C18 Columns address peak shape asymmetry and poor loading capacity for basic compounds in low ionic strength mobile phase. Surface charge facilitates fast column equilibration with low and high pH mobile phases. MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions, preventing poor peak shape and signal intensity losses. VanGuard FIT Column design protects from contaminants and extends lifetime.
  • Addresses peak shape asymmetry and poor loading capacity for basic compounds.
  • Facilitates fast column equilibration across pH ranges.
  • MaxPeak High Performance Surface reduces analyte/surface interactions.
  • VanGuard FIT Column design protects from contaminants and prolongs lifetime.
  • Highly efficient, offering wide selectivity and sensitivity.
Catalog No. 50-203-9070 Supplier Waters Corp Supplier No. 186009464
ACQUITY Premier CSH C18 Columns address peak shape asymmetry and poor loading capacity for basic compounds in low ionic strength mobile phase. Surface charge facilitates fast column equilibration with low and high pH mobile phases. MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions, preventing poor peak shape and signal intensity losses. VanGuard FIT Column design protects from contaminants and extends lifetime.
  • Addresses peak shape asymmetry and poor loading capacity for basic compounds.
  • Facilitates fast column equilibration across pH ranges.
  • MaxPeak High Performance Surface reduces analyte/surface interactions.
  • VanGuard FIT Column design protects from contaminants and prolongs lifetime.
  • Highly efficient, offering wide selectivity and sensitivity.