Learn More
Waters Corp Waters ACQUITY Premier CSH C18 Column, 1.7 μm, 2.1 mm, 100 mm
Supplier:  Waters Corp 186009461
Encompass_Preferred
The ACQUITY Premier CSH C18 Column is designed to overcome challenges in chromatographic separations, particularly with basic compounds. It features MaxPeak High Performance Surface hardware technology to minimize unwanted analyte/surface interactions, improving peak shape and signal intensity. This column offers enhanced analytical loadability and faster method development due to its CSH Technology.
  • MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • Provides increased analytical loadability, delivering sharper, narrower peaks for basic compounds.
Catalog No. 50-203-9067