Learn More
Waters Corp ACQUITY UPLC VanGuard FIT Cartridge, CSH Phenyl-Hexyl, 1.7 μm, 2.1 x 5 mm
Catalog No. 502039086 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY Premier cartridges work with the ACQUITY Premier VanGuard FIT columns. These fully integrated guard cartridges protect the analytical column from matrix and sample contaminants, extending column lifetime.
  • Phenyl chemistry
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range 1 to 11
  • Maximum pressure of 18000 psi (1240 bar)
  • Endcapped design
  • Phenyl-hexyl bonding technology
  • Low silanol activity
  • Spherical particle shape
  • Parker-style endfitting
  • Pore size of 130 Å
  • VanGuard FIT cartridge format
  • UPLC system compatibility
  • CSH particle technology
  • LC technique
  • USP classification L11
  • 14% carbon load
  • 3 units per package
Catalog No. 50-203-9086 Supplier Waters Corp Supplier No. 186009480
ACQUITY Premier cartridges work with the ACQUITY Premier VanGuard FIT columns. These fully integrated guard cartridges protect the analytical column from matrix and sample contaminants, extending column lifetime.
  • Phenyl chemistry
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range 1 to 11
  • Maximum pressure of 18000 psi (1240 bar)
  • Endcapped design
  • Phenyl-hexyl bonding technology
  • Low silanol activity
  • Spherical particle shape
  • Parker-style endfitting
  • Pore size of 130 Å
  • VanGuard FIT cartridge format
  • UPLC system compatibility
  • CSH particle technology
  • LC technique
  • USP classification L11
  • 14% carbon load
  • 3 units per package