Learn More
Waters Corp Column, CSH Phenyl-Hexyl, 1.7 μm, 2.1 x 100 mm
Catalog No. 502039084 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY Premier CSH Phenyl-Hexyl Columns provide alternative analyte selectivity and are valuable for method development. They feature a trifunctionally bonded C6 phenyl ligand for selective retention of polyaromatic compounds. MaxPeak High Performance Surface technology reduces unwanted analyte/surface interactions, and the VanGuard FIT Column design protects against contaminants, extending column lifetime.
  • Alternative analyte selectivity for method development.
  • Trifunctionally bonded C6 phenyl ligand retains polyaromatic compounds via pi-pi interactions.
  • MaxPeak High Performance Surface reduces analyte/surface interactions.
  • Integrated VanGuard FIT design protects against contaminants and extends column lifetime.
  • Facilitates UPLC method development.
  • Offers complementary selectivity to straight-chain alkyl phases.
  • Improves peak shape for basic compounds.
Catalog No. 50-203-9084 Supplier Waters Corp Supplier No. 186009478
ACQUITY Premier CSH Phenyl-Hexyl Columns provide alternative analyte selectivity and are valuable for method development. They feature a trifunctionally bonded C6 phenyl ligand for selective retention of polyaromatic compounds. MaxPeak High Performance Surface technology reduces unwanted analyte/surface interactions, and the VanGuard FIT Column design protects against contaminants, extending column lifetime.
  • Alternative analyte selectivity for method development.
  • Trifunctionally bonded C6 phenyl ligand retains polyaromatic compounds via pi-pi interactions.
  • MaxPeak High Performance Surface reduces analyte/surface interactions.
  • Integrated VanGuard FIT design protects against contaminants and extends column lifetime.
  • Facilitates UPLC method development.
  • Offers complementary selectivity to straight-chain alkyl phases.
  • Improves peak shape for basic compounds.