Learn More
Waters Corp Column, Phenyl, 1.7 μm, 2.1 x 150 mm
Catalog No. 502039082 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY CSH Phenyl-Hexyl Columns provide alternative analyte selectivity and are valuable for method development. They utilize a trifunctionally bonded C6 phenyl ligand, offering robust, low-bleed sorbents that selectively retain polyaromatic compounds via π–π interactions. MaxPeak High Performance Surface technology minimizes unwanted analyte/surface interactions, enhancing recovery, sensitivity, and reproducibility.
  • Minimizes unwanted analyte/surface interactions
  • Enhances analyte recovery, sensitivity, and reproducibility
  • Operates in a pH range of 1 to 11
  • Features trifunctionally bonded C6 phenyl ligand
  • Provides robust, low-bleed sorbents for polyaromatic compound retention
  • Reduces column conditioning and passivation times
  • Improves peak shapes and sensitivity
  • Allows for simpler mobile phases
  • Accelerates method development
Catalog No. 50-203-9082 Supplier Waters Corp Supplier No. 186009476
ACQUITY CSH Phenyl-Hexyl Columns provide alternative analyte selectivity and are valuable for method development. They utilize a trifunctionally bonded C6 phenyl ligand, offering robust, low-bleed sorbents that selectively retain polyaromatic compounds via π–π interactions. MaxPeak High Performance Surface technology minimizes unwanted analyte/surface interactions, enhancing recovery, sensitivity, and reproducibility.
  • Minimizes unwanted analyte/surface interactions
  • Enhances analyte recovery, sensitivity, and reproducibility
  • Operates in a pH range of 1 to 11
  • Features trifunctionally bonded C6 phenyl ligand
  • Provides robust, low-bleed sorbents for polyaromatic compound retention
  • Reduces column conditioning and passivation times
  • Improves peak shapes and sensitivity
  • Allows for simpler mobile phases
  • Accelerates method development