Learn More
Waters Corp Waters Column, Phenyl, 1.7 μm, 2.1 x 150 mm, VanGuard FIT
Supplier:  Waters Corp 186009479
Encompass_Preferred
ACQUITY Premier CSH Phenyl-Hexyl Columns are designed for method development, offering alternative analyte selectivity. They feature a robust C6 phenyl ligand, MaxPeak High Performance Surface hardware, and VanGuard FIT Column design for extended lifetime.
  • Provides alternative analyte selectivity and is a valuable tool for method development.
  • Trifunctionally bonded C6 phenyl ligand is a robust, low bleed sorbent that selectively retains polyaromatic compounds through pi-pi interactions.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions, which can lead to poor peak shape and losses in signal intensity.
  • VanGuard FIT Column design protects the column from contaminants and increases column lifetime.
Catalog No. 50-203-9085