Learn More
Waters Corp ACQUITY Method Validation Kit, CSH Phenyl-Hexyl, 1.7 μm, 130 Å, 2.1 x 100 mm
Catalog No. 502039124 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit offers alternative analyte selectivity for method development, featuring robust C6 phenyl ligand sorbents and MaxPeak High Performance Surface to prevent poor peak shape and signal loss.
  • Method Validation Kit with three columns (different material lots).
  • Robust, low bleed sorbents.
  • Selectively retains polyaromatic compounds via pi-pi interactions.
  • Particle size of 1.7 μm and pore size of 130 Å.
  • Designed for UPLC systems.
  • Maximum pressure of 18000 psi (1240 Bar).
Catalog No. 50-203-9124 Supplier Waters Corp Supplier No. 186009640
This method validation kit offers alternative analyte selectivity for method development, featuring robust C6 phenyl ligand sorbents and MaxPeak High Performance Surface to prevent poor peak shape and signal loss.
  • Method Validation Kit with three columns (different material lots).
  • Robust, low bleed sorbents.
  • Selectively retains polyaromatic compounds via pi-pi interactions.
  • Particle size of 1.7 μm and pore size of 130 Å.
  • Designed for UPLC systems.
  • Maximum pressure of 18000 psi (1240 Bar).