Learn More
Waters Corp Waters ACQUITY Premier CSH Phenyl-Hexyl Column, 1.7 μm, 2.1 mm, 50 mm
Catalog No. 502039080 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns offer alternative analyte selectivity, featuring a C6 phenyl ligand and MaxPeak High Performance Surface hardware for enhanced peak shape and signal intensity. Manufactured in a cGMP, ISO 9001-certified plant.
  • Provides alternative analyte selectivity.
  • Trifunctionally bonded C6 phenyl ligand acts as a robust, low bleed sorbent.
  • Selectively retains polyaromatic compounds through pi-pi interactions.
  • MaxPeak High Performance Surface hardware reduces unwanted analyte/surface interactions for improved peak shape and signal intensity.
  • Offers excellent peak shape for basic chemicals under acidic mobile-phase conditions.
  • Manufactured in a cGMP, ISO 9001-certified plant with ultra-pure reagents.
  • Independently analyzed with a performance chromatogram and batch analysis on the eCord intelligent chip.
Catalog No. 50-203-9080 Supplier Waters Corp Supplier No. 186009474
Add to Cart
Add to Cart
These columns offer alternative analyte selectivity, featuring a C6 phenyl ligand and MaxPeak High Performance Surface hardware for enhanced peak shape and signal intensity. Manufactured in a cGMP, ISO 9001-certified plant.
  • Provides alternative analyte selectivity.
  • Trifunctionally bonded C6 phenyl ligand acts as a robust, low bleed sorbent.
  • Selectively retains polyaromatic compounds through pi-pi interactions.
  • MaxPeak High Performance Surface hardware reduces unwanted analyte/surface interactions for improved peak shape and signal intensity.
  • Offers excellent peak shape for basic chemicals under acidic mobile-phase conditions.
  • Manufactured in a cGMP, ISO 9001-certified plant with ultra-pure reagents.
  • Independently analyzed with a performance chromatogram and batch analysis on the eCord intelligent chip.