Learn More
Waters Corp Waters Column, Glycan BEH Amide, 130Å, 1.7 μm, 2.1 × 150 mm
Supplier:  Waters Corp 186009524
Encompass_Preferred
This column is ideal for HILIC-mode separations of labeled glycans, including 2-aminobenzamide (2-AB), 2-aminobenzoic acid (2-AA), or RapiFluor-MS (RFMS). It ensures batch-to-batch consistency through rigorous quality control testing. The column features MaxPeak High Performance Surface (HPS) for improved peak shape, recovery, and reproducibility by reducing unwanted analyte surface interactions.
  • Small particle size enables high resolution in complex sample matrices.
  • Not susceptible to Schiff base formation, improving quantitation accuracy.
  • Trifunctionally-bonded amide phase ensures repeatable results across varying experiments.
  • Useful for retaining polar analytes that are too polar for reverse-phase chromatography.
  • Offers orthogonal selectivity compared to reversed-phase materials.
Catalog No. 50-222-4229