Learn More
Waters Corp ACQUITY UPLC Column, Glycoprotein BEH Amide, 300Å, 1.7 μm, 2.1 x 50 mm
Supplier:  Waters Corp 186009547
Encompass_Preferred
ACQUITY Premier Glycoprotein BEH Amide 300Å, 1.7 μm, 2.1 x 50 mm Columns are designed for analysis of intact glycoproteins, fragments, and glycopeptides. Material is quality control tested with the Glycoprotein Performance Test Standard for batch consistency. Column hardware utilizes MaxPeak High Performance Surface (HPS) to reduce unwanted analyte interactions, improving peak shape, recovery, and reproducibility.
  • Professionally tested with glycoprotein performance test standard to ensure premium performance.
  • MaxPeak High Performance Surface Technology to greatly reduce unwanted analyte-surface interactions.
  • Sub-2μm particle technology maximizes separation performance.
  • Incorporates easily with HILIC method development, aiding in MS adoption.
  • Works well with UPLC systems.
Catalog No. 50-222-4230