Learn More
Waters Corp Waters ACQUITY Premier Glycoprotein BEH Amide Column, 300Å, 1.7 μm, 2.1 x 150 mm
Catalog No. 502224232 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These ACQUITY Premier Glycoprotein BEH Amide columns are designed for analyzing intact glycoproteins, fragments, and glycopeptides. Each batch undergoes quality control testing for consistency. MaxPeak High Performance Surface (HPS) technology minimizes unwanted analyte interactions, enhancing peak shape, recovery, and reproducibility.
  • Enables HILIC method development for intact proteins.
  • Reduces sample loss; requires no conditioning, saving time and reagents.
  • Increases retention time, peak response, and throughput.
  • Optimized for UPLC, handling wide flow rates and mobile phase extremes.
  • Utilizes a stable, trifunctionally-bonded amide phase for efficient HILIC separations.
Catalog No. 50-222-4232 Supplier Waters Corp Supplier No. 186009549
These ACQUITY Premier Glycoprotein BEH Amide columns are designed for analyzing intact glycoproteins, fragments, and glycopeptides. Each batch undergoes quality control testing for consistency. MaxPeak High Performance Surface (HPS) technology minimizes unwanted analyte interactions, enhancing peak shape, recovery, and reproducibility.
  • Enables HILIC method development for intact proteins.
  • Reduces sample loss; requires no conditioning, saving time and reagents.
  • Increases retention time, peak response, and throughput.
  • Optimized for UPLC, handling wide flow rates and mobile phase extremes.
  • Utilizes a stable, trifunctionally-bonded amide phase for efficient HILIC separations.