Learn More
Waters Corp Waters Glycoprotein Column, 1.7 μm, 300Å, 2.1 x 100 mm
Supplier:  Waters Corp 186009548
Encompass_Preferred
The ACQUITY Premier Glycoprotein BEH Amide 300Å, 1.7 μm, 2.1 x 100 mm columns are designed for the analysis of intact glycoproteins, glycoprotein fragments, and glycopeptides. The column hardware utilizes MaxPeak High Performance Surface (HPS) that reduces unwanted analyte surface interactions and greatly improves peak shape, recovery, and reproducibility.
  • Reduce unwanted analyte surface interactions and improve peak shape, recovery, and reproducibility using MaxPeak High Performance Surface (HPS).
  • Specifically quality control tested with a Glycoprotein Performance Test Standard for batch to batch consistency.
  • Designed for analysis of intact glycoproteins, glycoprotein fragments, and glycopeptides.
Catalog No. 50-222-4231