Learn More
Waters Corp Waters Column, Amide, 1.7 μm, 300 Å, 2.1 mm x 100 mm
Supplier:  Waters Corp 176004867
Encompass_Preferred
ACQUITY Premier Glycoprotein BEH Amide Columns are for analyzing intact glycoproteins, fragments, and glycopeptides. They feature MaxPeak HPS to improve peak shape, recovery, and reproducibility. A Glycoprotein Performance Test Standard is included for benchmarking.
  • Designed for analysis of intact glycoproteins, fragments, and glycopeptides.
  • Batch quality control tested with Glycoprotein Performance Test Standard for consistency.
  • MaxPeak High Performance Surface (HPS) improves peak shape, recovery, and reproducibility by reducing unwanted analyte interactions.
  • Includes one PREMIER Glycoprotein BEH Amide column and one Glycoprotein Performance Test Standard.
  • Standard for immediate benchmarking upon receipt and continuous monitoring.
Catalog No. 50-222-4153