Learn More
Waters Corp ACQUITY Premier HSS C18 Column, 1.8 μm, 2.1 mm, 150 mm
Supplier:  Waters Corp 186010017
Encompass_Preferred
These columns feature an ultra-performance general-purpose C18 bonded phase, providing increased retention. They utilize high strength silica (HSS) particles and a tri-functional C18 bonded phase for extended column lifetimes at low pH. The columns also incorporate MaxPeak High Performance Surface hardware technology to minimize unwanted analyte/surface interactions, which helps prevent poor peak shape and reduced signal intensity.
  • Increased retention due to general purpose C18 bonded phase.
  • Extremely long column lifetimes at low pH, provided by high strength silica (HSS) particles and tri-functional C18 bonded phase.
  • Reduced unwanted analyte/surface interactions with MaxPeak High Performance Surface hardware technology.
  • Improved peak shape and signal intensity due to reduced analyte/surface interactions.
Catalog No. 50-239-1902