Learn More
Waters Corp Column, C18, 1.8 μm, 2.1 mm x 50 mm
Catalog No. 502391707 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These HSS C18 columns offer increased retention with an ultra-performance C18 bonded phase. High strength silica (HSS) particles and a tri-functional C18 bonded phase ensure extended column lifetimes, especially at low pH. MaxPeak High Performance Surface hardware minimizes unwanted analyte/surface interactions, improving peak shape and signal intensity. A VanGuard Fit design protects against contaminants, extending lifespan.
  • Ultra-performance general purpose C18 bonded phase provides increased retention.
  • High strength silica (HSS) particle and tri-functional C18 bonded phase contribute to extremely long column lifetimes at low pH.
  • MaxPeak High Performance Surface hardware significantly reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • VanGuard Fit column design protects the column from contaminants and increases column lifetime.
Catalog No. 50-239-1707 Supplier Waters Corp Supplier No. 186010018
These HSS C18 columns offer increased retention with an ultra-performance C18 bonded phase. High strength silica (HSS) particles and a tri-functional C18 bonded phase ensure extended column lifetimes, especially at low pH. MaxPeak High Performance Surface hardware minimizes unwanted analyte/surface interactions, improving peak shape and signal intensity. A VanGuard Fit design protects against contaminants, extending lifespan.
  • Ultra-performance general purpose C18 bonded phase provides increased retention.
  • High strength silica (HSS) particle and tri-functional C18 bonded phase contribute to extremely long column lifetimes at low pH.
  • MaxPeak High Performance Surface hardware significantly reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • VanGuard Fit column design protects the column from contaminants and increases column lifetime.