Learn More
Waters Corp Column, HSS PFP, 1.8 μm, 2.1 mm x 50 mm
Catalog No. 502390809 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY Premier HSS PFP Column is a non-endcapped silica-based column for low pH separations, offering alternative selectivity to C18 phases. It uses MaxPeak High Performance Surface technology to reduce analyte/surface interactions, improving peak shape and signal intensity.
  • Non-endcapped silica-based column for low pH separations.
  • Increased retentivity over hybrid materials.
  • Offers widest selectivity space with C18, T3, C18 SB, Cyano, PFP chemistries.
  • High strength silica provides mechanical stability.
  • Reduced column conditioning and passivation times.
  • Improved sensitivity and peak shapes.
  • Improved recovery.
  • Simpler mobile phases, without complex additives.
  • Time savings in method development.
  • Reduced risk, greater data confidence, and decision making.
Catalog No. 50-239-0809 Supplier Waters Corp Supplier No. 186010036
The ACQUITY Premier HSS PFP Column is a non-endcapped silica-based column for low pH separations, offering alternative selectivity to C18 phases. It uses MaxPeak High Performance Surface technology to reduce analyte/surface interactions, improving peak shape and signal intensity.
  • Non-endcapped silica-based column for low pH separations.
  • Increased retentivity over hybrid materials.
  • Offers widest selectivity space with C18, T3, C18 SB, Cyano, PFP chemistries.
  • High strength silica provides mechanical stability.
  • Reduced column conditioning and passivation times.
  • Improved sensitivity and peak shapes.
  • Improved recovery.
  • Simpler mobile phases, without complex additives.
  • Time savings in method development.
  • Reduced risk, greater data confidence, and decision making.