Learn More
Waters Corp ACQUITY UPLC Column, 1.8 μm, 2.1 mm x 150 mm, PFP/Fluoro-Phenyl
Catalog No. 502390712 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY Premier HSS PFP Column is a non-endcapped silica-based column for low pH separations, offering alternative selectivity to C18 phases. It features MaxPeak High Performance Surface hardware technology, significantly reducing unwanted analyte/surface interactions to improve peak shape, signal intensity, analyte recovery, and reproducibility.
  • Reduces unwanted analyte/surface interactions, mitigating loss of metal-sensitive analytes.
  • Increases analyte recovery and reproducibility.
  • Improves peak shape and sensitivity for accurate analysis.
  • Supports enhanced low-level detection, for instance, in peptide analysis.
  • Allows for simpler mobile phases without complex additives.
  • Reduces column conditioning and passivation times.
Catalog No. 50-239-0712 Supplier Waters Corp Supplier No. 186010038
The ACQUITY Premier HSS PFP Column is a non-endcapped silica-based column for low pH separations, offering alternative selectivity to C18 phases. It features MaxPeak High Performance Surface hardware technology, significantly reducing unwanted analyte/surface interactions to improve peak shape, signal intensity, analyte recovery, and reproducibility.
  • Reduces unwanted analyte/surface interactions, mitigating loss of metal-sensitive analytes.
  • Increases analyte recovery and reproducibility.
  • Improves peak shape and sensitivity for accurate analysis.
  • Supports enhanced low-level detection, for instance, in peptide analysis.
  • Allows for simpler mobile phases without complex additives.
  • Reduces column conditioning and passivation times.