Learn More
Waters Corp VanGuard FIT Cartridge, PFP, HSS, 1.8 μm
Supplier:  Waters Corp 186010042
Encompass_Preferred
ACQUITY Premier Cartridges are designed to work seamlessly with ACQUITY Premier VanGuard FIT columns. These integrated guard cartridges serve to protect the analytical column from contaminants present in the matrix and sample, thereby prolonging the column's lifetime.
  • The cartridges protect the analytical column from matrix and sample contaminants.
  • They help extend the lifetime of the analytical column.
  • They are fully integrated with compatible columns.
Catalog No. 50-239-0718