Learn More
Waters Corp ACQUITY Premier HSS T3 Column, 100 Å, 1.8 μm, 2.1 x 50 mm
Catalog No. 502391565 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit includes three columns with different material lots, utilizing universal, silica-based T3 sorbents for diverse compound separations. They offer ultra-low MS bleed, are scalable to HPLC, and integrate MaxPeak HPS technology with VanGuard Fit design for optimized performance and longevity.
  • Compatible with 100% aqueous mobile phase for varied applications.
  • Suitable for developing separations of both polar and non-polar compounds.
  • Characterized by ultra-low MS bleed.
  • Fully scalable and transferable to HPLC separations.
  • MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions.
  • VanGuard Fit column design protects the column from contaminants.
  • Extends operational lifetime.
Catalog No. 50-239-1565 Supplier Waters Corp Supplier No. 186009636
This method validation kit includes three columns with different material lots, utilizing universal, silica-based T3 sorbents for diverse compound separations. They offer ultra-low MS bleed, are scalable to HPLC, and integrate MaxPeak HPS technology with VanGuard Fit design for optimized performance and longevity.
  • Compatible with 100% aqueous mobile phase for varied applications.
  • Suitable for developing separations of both polar and non-polar compounds.
  • Characterized by ultra-low MS bleed.
  • Fully scalable and transferable to HPLC separations.
  • MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions.
  • VanGuard Fit column design protects the column from contaminants.
  • Extends operational lifetime.