Learn More
Waters Corp ACQUITY Column Kit, C18, 1.8 μm, 100 Å, 2.1 x 100 mm, 3 pk
Catalog No. 502391550 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY Premier HSS T3 Column with VanGuard Fit is a method validation kit (MVK) containing three columns, each packed with a different lot of material.
  • Universal, silica-based bonded phase compatible with 100% aqueous mobile phase, suitable for developing separations of polar and non-polar compounds.
  • Exhibits ultra-low MS bleed, beneficial for mass spectrometry applications.
  • Fully scalable and transferable to HPLC separations.
  • MaxPeak high performance surface hardware technology significantly reduces unwanted analyte/surface interactions, preventing poor peak shape and signal intensity loss.
  • VanGuard Fit column design protects the column from contaminants and increases its lifetime.
Catalog No. 50-239-1550 Supplier Waters Corp Supplier No. 186009637
The ACQUITY Premier HSS T3 Column with VanGuard Fit is a method validation kit (MVK) containing three columns, each packed with a different lot of material.
  • Universal, silica-based bonded phase compatible with 100% aqueous mobile phase, suitable for developing separations of polar and non-polar compounds.
  • Exhibits ultra-low MS bleed, beneficial for mass spectrometry applications.
  • Fully scalable and transferable to HPLC separations.
  • MaxPeak high performance surface hardware technology significantly reduces unwanted analyte/surface interactions, preventing poor peak shape and signal intensity loss.
  • VanGuard Fit column design protects the column from contaminants and increases its lifetime.