Learn More
Waters Corp Waters Column Kit, 1.8 μm, 100 Å, 2.1 x 100 mm
Catalog No. 502039118 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit contains three columns for developing separations of polar and non-polar compounds. Featuring a universal, silica-based bonded phase compatible with 100% aqueous mobile phase, these columns provide ultra-low MS bleed and are fully scalable to HPLC. MaxPeak High Performance Surface hardware technology improves performance by reducing unwanted analyte/surface interactions.
  • Method validation kit (MVK) includes three columns, each from a different material lot.
  • Universal, silica-based bonded phase compatible with 100% aqueous mobile phase, ideal for polar and non-polar compound separations.
  • Exhibits ultra-low MS bleed.
  • Fully scalable and transferable to HPLC separations.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
Catalog No. 50-203-9118 Supplier Waters Corp Supplier No. 186009634
This method validation kit contains three columns for developing separations of polar and non-polar compounds. Featuring a universal, silica-based bonded phase compatible with 100% aqueous mobile phase, these columns provide ultra-low MS bleed and are fully scalable to HPLC. MaxPeak High Performance Surface hardware technology improves performance by reducing unwanted analyte/surface interactions.
  • Method validation kit (MVK) includes three columns, each from a different material lot.
  • Universal, silica-based bonded phase compatible with 100% aqueous mobile phase, ideal for polar and non-polar compound separations.
  • Exhibits ultra-low MS bleed.
  • Fully scalable and transferable to HPLC separations.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.