Learn More
Waters Corp Waters ACQUITY Premier HSS T3 Column, 1.8 μm, 2.1 mm ID, 50 mm Length
Catalog No. 502039073 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY Premier HSS T3 Column is a universal, silica-based bonded phase column compatible with 100% aqueous mobile phases for polar and non-polar compounds. It is manufactured in a cGMP, ISO 9001 certified plant, with each batch tested for reproducible performance.
  • Universal silica-based bonded phase, compatible with 100% aqueous mobile phases for polar and non-polar compound separation.
  • Exhibits ultra-low MS bleed.
  • Scalable and transferable to HPLC separations.
  • MaxPeak High Performance Surface (HPS) technology reduces analyte/surface interactions, improving peak shape, signal, recovery, sensitivity, and reproducibility.
  • HSS (High Strength Silica) particle technology withstands UPLC pressures, offering mechanically tolerant, silica-based material for increased retentivity and alternate selectivity.
  • Operates within a 2 to 8 pH range.
Catalog No. 50-203-9073 Supplier Waters Corp Supplier No. 186009467
The ACQUITY Premier HSS T3 Column is a universal, silica-based bonded phase column compatible with 100% aqueous mobile phases for polar and non-polar compounds. It is manufactured in a cGMP, ISO 9001 certified plant, with each batch tested for reproducible performance.
  • Universal silica-based bonded phase, compatible with 100% aqueous mobile phases for polar and non-polar compound separation.
  • Exhibits ultra-low MS bleed.
  • Scalable and transferable to HPLC separations.
  • MaxPeak High Performance Surface (HPS) technology reduces analyte/surface interactions, improving peak shape, signal, recovery, sensitivity, and reproducibility.
  • HSS (High Strength Silica) particle technology withstands UPLC pressures, offering mechanically tolerant, silica-based material for increased retentivity and alternate selectivity.
  • Operates within a 2 to 8 pH range.