Learn More
Waters Corp ACQUITY Premier HSS T3 Column, 1.8 μm, 2.1 x 50 mm
Catalog No. 502390571 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column features a universal, silica-based bonded phase compatible with 100% aqueous mobile phase, suitable for developing separations for polar and non-polar compounds. It offers ultra-low MS bleed and is fully scalable to HPLC. MaxPeak High Performance Surface hardware reduces unwanted analyte/surface interactions, preventing poor peak shape and signal losses. The VanGuard FIT Column design protects the column from contaminants, extending its lifetime.
  • Maximizes UPLC column efficiency, aids MS adoption.
  • High-Strength Silica (HSS) particle technology increases retentivity of polar and non-polar compounds.
  • Provides increased mechanical stability under higher backpressures.
  • VanGuard FIT column protection prevents signal intensity losses from fouling.
Catalog No. 50-239-0571 Supplier Waters Corp Supplier No. 186009470
This column features a universal, silica-based bonded phase compatible with 100% aqueous mobile phase, suitable for developing separations for polar and non-polar compounds. It offers ultra-low MS bleed and is fully scalable to HPLC. MaxPeak High Performance Surface hardware reduces unwanted analyte/surface interactions, preventing poor peak shape and signal losses. The VanGuard FIT Column design protects the column from contaminants, extending its lifetime.
  • Maximizes UPLC column efficiency, aids MS adoption.
  • High-Strength Silica (HSS) particle technology increases retentivity of polar and non-polar compounds.
  • Provides increased mechanical stability under higher backpressures.
  • VanGuard FIT column protection prevents signal intensity losses from fouling.