Learn More
Waters Corp Waters ACQUITY Premier HSS T3 Column, 1.8 μm, 2.1 mm, 100 mm
Catalog No. 502039074 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This ACQUITY Premier HSS T3 UPLC Column offers a universal, silica-based bonded phase compatible with 100% aqueous mobile phases, ideal for separating polar and non-polar compounds. It features ultra-low MS bleed, is scalable to HPLC, and uses MaxPeak High Performance Surface hardware to minimize unwanted analyte interactions.
  • Universal silica-based bonded phase, 100% aqueous mobile phase compatible.
  • Separates polar and non-polar compounds.
  • Ultra-low MS bleed.
  • Scalable and transferable to HPLC.
  • MaxPeak High Performance Surface hardware reduces unwanted analyte interactions.
  • Low ligand density enhances retention of polar chemicals/metabolites.
  • HSS particle technology offers mechanical stability under UPLC pressures.
  • Improved retentivity and alternative selectivity versus hybrid particle technology.
  • 2 to 8 pH range.
Catalog No. 50-203-9074 Supplier Waters Corp Supplier No. 186009468
Add to Cart
Add to Cart
This ACQUITY Premier HSS T3 UPLC Column offers a universal, silica-based bonded phase compatible with 100% aqueous mobile phases, ideal for separating polar and non-polar compounds. It features ultra-low MS bleed, is scalable to HPLC, and uses MaxPeak High Performance Surface hardware to minimize unwanted analyte interactions.
  • Universal silica-based bonded phase, 100% aqueous mobile phase compatible.
  • Separates polar and non-polar compounds.
  • Ultra-low MS bleed.
  • Scalable and transferable to HPLC.
  • MaxPeak High Performance Surface hardware reduces unwanted analyte interactions.
  • Low ligand density enhances retention of polar chemicals/metabolites.
  • HSS particle technology offers mechanical stability under UPLC pressures.
  • Improved retentivity and alternative selectivity versus hybrid particle technology.
  • 2 to 8 pH range.