Learn More
Waters Corp ACQUITY Premier Oligonucleotide C18 Column, 130 Å, 1.7 μm, 2.1 mm x 150 mm
Catalog No. 502390715 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY Premier Oligonucleotide BEH C18 Columns separate synthetic oligonucleotides via ion-pairing reversed-phase (IP-RP) chromatography. QC tested with Waters MassPrep OST standards for repeatable performance. These MaxPeak Premier columns feature an ultra-low non-adsorptive surface, enhancing sensitivity and reproducibility for metal-sensitive analytes.
  • Separates synthetic oligonucleotides via ion-pairing reversed-phase chromatography.
  • QC tested with MassPrep OST standards for repeatable performance.
  • MaxPeak Premier technology for ultra-low non-adsorptive surface.
  • Maximizes sensitivity and reproducibility for metal-sensitive analytes.
  • Integrated, replaceable 5 mm pre-column extends analytical column life.
  • Pre-column protects without dispersion or impacting resolution.
  • VanGuard FIT Cartridge replacements use same qualified sorbent.
Catalog No. 50-239-0715 Supplier Waters Corp Supplier No. 186010687
ACQUITY Premier Oligonucleotide BEH C18 Columns separate synthetic oligonucleotides via ion-pairing reversed-phase (IP-RP) chromatography. QC tested with Waters MassPrep OST standards for repeatable performance. These MaxPeak Premier columns feature an ultra-low non-adsorptive surface, enhancing sensitivity and reproducibility for metal-sensitive analytes.
  • Separates synthetic oligonucleotides via ion-pairing reversed-phase chromatography.
  • QC tested with MassPrep OST standards for repeatable performance.
  • MaxPeak Premier technology for ultra-low non-adsorptive surface.
  • Maximizes sensitivity and reproducibility for metal-sensitive analytes.
  • Integrated, replaceable 5 mm pre-column extends analytical column life.
  • Pre-column protects without dispersion or impacting resolution.
  • VanGuard FIT Cartridge replacements use same qualified sorbent.