Learn More
Waters Corp Column, C18, 130 Å, 1.7 μm, 2.1 x 50 mm
Catalog No. 502391977 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns are designed for the separation of synthetic oligonucleotides using ion-pairing reversed-phase (IP-RP) chromatography, ensuring consistent performance for metal-sensitive analytes.
  • Separates synthetic oligonucleotides based on ion-pairing reversed-phase (IP-RP) chromatography.
  • QC tested using MassPrep OST standards for repeatable performance.
  • MaxPeak Premier line delivers an ultra-low non-adsorptive surface, maximizing sensitivity and reproducibility for metal-sensitive analytes.
  • Includes a fully integrated, easily replaceable 5 mm pre-column to protect and extend analytical column life without impacting resolution.
  • VanGuard FIT Cartridge replacements contain the same qualified sorbent and are easily replaceable.
Catalog No. 50-239-1977 Supplier Waters Corp Supplier No. 186010685
Add to Cart
Add to Cart
These columns are designed for the separation of synthetic oligonucleotides using ion-pairing reversed-phase (IP-RP) chromatography, ensuring consistent performance for metal-sensitive analytes.
  • Separates synthetic oligonucleotides based on ion-pairing reversed-phase (IP-RP) chromatography.
  • QC tested using MassPrep OST standards for repeatable performance.
  • MaxPeak Premier line delivers an ultra-low non-adsorptive surface, maximizing sensitivity and reproducibility for metal-sensitive analytes.
  • Includes a fully integrated, easily replaceable 5 mm pre-column to protect and extend analytical column life without impacting resolution.
  • VanGuard FIT Cartridge replacements contain the same qualified sorbent and are easily replaceable.