Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp ACQUITY Oligonucleotide Column, C18, 1.7 μm, 2.1 x 150 mm

Catalog No. 502392823 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These Premier Oligonucleotide columns are designed for the separation of detritylated synthetic oligonucleotides using ion-pair, reversed-phase chromatography. They offer enhanced reproducibility, improved peak shape, and better recovery for metal-sensitive analytes. The method validation kit includes three columns, each packed with a different lot of material.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Hybrid
  • Particle size: 1.7 μm
  • Pore size: 130 Å
  • Inner diameter: 2.1 mm
  • Length: 150 mm
  • Maximum pressure: 18000 psi (1240 Bar)
  • pH range: 1 to 12 pH
  • Format: Method validation kit
  • Units per package: 3 pk
  • Application: Oligonucleotide, gene therapeutics

Catalog No. 50-239-2823 Supplier Waters Corp Supplier No. 186009650
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

These Premier Oligonucleotide columns are designed for the separation of detritylated synthetic oligonucleotides using ion-pair, reversed-phase chromatography. They offer enhanced reproducibility, improved peak shape, and better recovery for metal-sensitive analytes. The method validation kit includes three columns, each packed with a different lot of material.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Hybrid
  • Particle size: 1.7 μm
  • Pore size: 130 Å
  • Inner diameter: 2.1 mm
  • Length: 150 mm
  • Maximum pressure: 18000 psi (1240 Bar)
  • pH range: 1 to 12 pH
  • Format: Method validation kit
  • Units per package: 3 pk
  • Application: Oligonucleotide, gene therapeutics

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.