Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Column Kit, C18, 130Å, 1.7 μm, 2.1 x 50 mm, MVK

Catalog No. 502039132 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These oligonucleotide columns are for separating detritylated synthetic oligonucleotides via ion-pair, reversed-phase chromatography. QC tested for consistent performance in demanding assays, they are part of the Premier class, enhancing reproducibility, peak shape, and recovery for metal-sensitive analytes. This method validation kit (MVK) includes three columns.

  • C18 chemistry
  • Reversed phase separation
  • pH range of 1 to 12
  • Maximum pressure up to 18000 psi (1240 Bar)
  • Endcapped design
  • Particle size: 1.7 μm
  • Pore size: 130 Å
  • QC tested for oligonucleotide applications
  • Method validation kit format
  • UPLC system compatible
  • Inner diameter: 2.1 mm
  • Length: 50 mm
  • Application for oligonucleotide and gene therapeutics
  • 3 units per package

Catalog No. 50-203-9132 Supplier Waters Corp Supplier No. 186009648
May include imposed supplier surcharges.
Add to Cart
Add to Cart

These oligonucleotide columns are for separating detritylated synthetic oligonucleotides via ion-pair, reversed-phase chromatography. QC tested for consistent performance in demanding assays, they are part of the Premier class, enhancing reproducibility, peak shape, and recovery for metal-sensitive analytes. This method validation kit (MVK) includes three columns.

  • C18 chemistry
  • Reversed phase separation
  • pH range of 1 to 12
  • Maximum pressure up to 18000 psi (1240 Bar)
  • Endcapped design
  • Particle size: 1.7 μm
  • Pore size: 130 Å
  • QC tested for oligonucleotide applications
  • Method validation kit format
  • UPLC system compatible
  • Inner diameter: 2.1 mm
  • Length: 50 mm
  • Application for oligonucleotide and gene therapeutics
  • 3 units per package

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.