Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Column, C18, 1.7 μm, 2.1 x 150 mm, 130 Å

Catalog No. 502039089 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

ACQUITY Premier Oligonucleotide Columns are designed for ion-pair, reversed-phase chromatography of detritylated synthetic oligonucleotides. They provide enhanced reproducibility, peak shape, and recovery for metal-sensitive analytes, utilizing BEH particle technology for method development efficiency and stability.

  • Chemistry: C18
  • Separation mode: Reversed Phase
  • Particle substrate: Hybrid
  • pH range: 1 to 12
  • Maximum pressure: 18000 psi (1240 Bar)
  • Particle size: 1.7 μm
  • Pore size: 130 Å
  • QC tested: Oligonucleotide
  • System: UPLC
  • Particle technology: BEH
  • USP classification: L1
  • Inner diameter: 2.1 mm
  • Length: 150 mm
  • Carbon load: 18%
  • Application: Oligonucleotide, Gene Therapeutics

Catalog No. 50-203-9089 Supplier Waters Corp Supplier No. 186009486
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACQUITY Premier Oligonucleotide Columns are designed for ion-pair, reversed-phase chromatography of detritylated synthetic oligonucleotides. They provide enhanced reproducibility, peak shape, and recovery for metal-sensitive analytes, utilizing BEH particle technology for method development efficiency and stability.

  • Chemistry: C18
  • Separation mode: Reversed Phase
  • Particle substrate: Hybrid
  • pH range: 1 to 12
  • Maximum pressure: 18000 psi (1240 Bar)
  • Particle size: 1.7 μm
  • Pore size: 130 Å
  • QC tested: Oligonucleotide
  • System: UPLC
  • Particle technology: BEH
  • USP classification: L1
  • Inner diameter: 2.1 mm
  • Length: 150 mm
  • Carbon load: 18%
  • Application: Oligonucleotide, Gene Therapeutics

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.