Learn More
Waters Corp ACQUITY Premier Peptide BEH C18 Column, 300Å, 1.7 μm
Catalog No. 502391535 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This ACQUITY Premier Peptide BEH C18 Column features 300Å pore and 1.7 μm particle size, optimized for peptide separations, and includes three columns in a method validation kit.
  • Optimized for peptide separations.
  • Wide-pore (300Å) BEH particle.
  • Wide pH range (1-12) & stable at low pH.
  • Ultra-low column bleed.
  • Ideal for large peptides (proteomics/protein characterization).
  • For UPLC/UHPLC & LC-MS methods.
  • Enhances reproducibility & recovery of metal-sensitive analytes.
  • Method validation kit (3 columns with different lot materials).
Catalog No. 50-239-1535 Supplier Waters Corp Supplier No. 186009646
Add to Cart
Add to Cart
Request a Quote
This ACQUITY Premier Peptide BEH C18 Column features 300Å pore and 1.7 μm particle size, optimized for peptide separations, and includes three columns in a method validation kit.
  • Optimized for peptide separations.
  • Wide-pore (300Å) BEH particle.
  • Wide pH range (1-12) & stable at low pH.
  • Ultra-low column bleed.
  • Ideal for large peptides (proteomics/protein characterization).
  • For UPLC/UHPLC & LC-MS methods.
  • Enhances reproducibility & recovery of metal-sensitive analytes.
  • Method validation kit (3 columns with different lot materials).