Learn More
Waters Corp Waters Column, ACQUITY Premier Peptide BEH C18, 300 Angstrom, 1.7 Micrometer, 2.1 x 100 mm
Supplier:  Waters Corp 186009494
Encompass_Preferred
The ACQUITY Premier Peptide BEH C18 Column is optimized for peptide separations, utilizing wide-pore (300Å) trifunctionally bonded BEH particles to offer a wide pH range, superior low pH stability, and ultra-low column bleed. It enhances reproducibility, peak shape, and recovery for metal-sensitive analytes in UPLC/UHPLC and LC-MS applications.
  • Optimized and QC tested for peptide separations
  • Wide-pore (300Å) trifunctionally bonded BEH particle
  • Wide usable pH range (1 to 12)
  • Superior low pH stability
  • Ultra-low column bleed
  • Recommended for UPLC or UHPLC systems
  • Enhances reproducibility, peak shape, and recovery for metal-sensitive analytes
  • C18 chemistry
  • 1.7 μm particle size
  • 2.1 mm inner diameter, 100 mm length
  • Maximum operating temperature of 80°C
  • Maximum pressure of 18000 psi (1240 Bar)
Catalog No. 50-203-9095